Product Details

SNP ID
rs78568470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38201694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGTATCATGTCTTTACCTTC[C/T]GGCTACCTCCTTGGGTTAGCCTTGC
Phenotype
MIM: 603884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAG4 PubMed Links

Gene Details

Gene
BAG4
Gene Name
BCL2 associated athanogene 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204878.1 Intron NP_001191807.1
NM_004874.3 Intron NP_004865.1

View Full Product Details