Product Details

SNP ID
rs77891975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:12724780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAATCAGATTCAAGTAATCAG[G/T]TAAAACAAATAGATGGCAACTGATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LONRF1 PubMed Links

Gene Details

Gene
LONRF1
Gene Name
LON peptidase N-terminal domain and ring finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005273685.3 Intron XP_005273742.1
XM_011544694.2 Intron XP_011542996.1
Gene
MIR3926-1
Gene Name
microRNA 3926-1
There are no transcripts associated with this gene.

Gene
MIR3926-2
Gene Name
microRNA 3926-2
There are no transcripts associated with this gene.

Gene
MIR5692A2
Gene Name
microRNA 5692a-2
There are no transcripts associated with this gene.

View Full Product Details