Product Details

SNP ID
rs75337766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22040717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGAGTCCCGACAGCATAAATT[G/T]GAACCGTTACCCACCTTTCGCCCCC
Phenotype
MIM: 125305 MIM: 603725 MIM: 608073
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DMTN PubMed Links

Gene Details

Gene
DMTN
Gene Name
dematin actin binding protein
There are no transcripts associated with this gene.

Gene
FGF17
Gene Name
fibroblast growth factor 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304478.1 714 Intron NP_001291407.1
NM_003867.3 714 Intron NP_003858.1
XM_005273675.1 714 Intron XP_005273732.1
XM_011544683.1 714 UTR 5 XP_011542985.1
XM_011544684.2 714 Intron XP_011542986.1
XM_011544685.1 714 UTR 5 XP_011542987.1
Gene
NPM2
Gene Name
nucleophosmin/nucleoplasmin 2
There are no transcripts associated with this gene.

View Full Product Details