Product Details

SNP ID
rs75933844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65640492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCAATACAAGTCATCTGTCC[A/G]GTTGGGCTAGGTCCTGGTGGGACCC
Phenotype
MIM: 602180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4489 PubMed Links

Gene Details

Gene
MIR4489
Gene Name
microRNA 4489
There are no transcripts associated with this gene.

Gene
MIR4690
Gene Name
microRNA 4690
There are no transcripts associated with this gene.

Gene
PCNX3
Gene Name
pecanex homolog 3 (Drosophila)
There are no transcripts associated with this gene.

Gene
SIPA1
Gene Name
signal-induced proliferation-associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006747.3 Intron NP_006738.3
NM_153253.29 Intron NP_694985.29
XM_005274189.2 Intron XP_005274246.1
XM_011545214.1 Intron XP_011543516.1

View Full Product Details