Product Details

SNP ID
rs74853303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72220969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAACTTTAACTGGGACCACTGC[A/G]GCAAGATGGAGCCCGCCTGCAAGCG
Phenotype
MIM: 136425 MIM: 600829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLR2 PubMed Links

Gene Details

Gene
FOLR2
Gene Name
folate receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000803.4 407 Missense Mutation AGC,GGC S84G NP_000794.3
NM_001113534.1 407 Missense Mutation AGC,GGC S84G NP_001107006.1
NM_001113535.1 407 Missense Mutation AGC,GGC S84G NP_001107007.1
NM_001113536.1 407 Missense Mutation AGC,GGC S84G NP_001107008.1
XM_005273856.3 407 Missense Mutation AGC,GGC S93G XP_005273913.1
Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
There are no transcripts associated with this gene.

View Full Product Details