Product Details

SNP ID
rs79247300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73269511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAAAATAAGGCCTGTGGACTA[A/G]GGGTTTGCGGGGCTCCCCAGGAGCA
Phenotype
MIM: 602451
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P2RY6 PubMed Links

Gene Details

Gene
P2RY6
Gene Name
pyrimidinergic receptor P2Y6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277204.1 909 Intron NP_001264133.1
NM_001277205.1 909 Intron NP_001264134.1
NM_001277206.1 909 Intron NP_001264135.1
NM_001277207.1 909 Intron NP_001264136.1
NM_001277208.1 909 Intron NP_001264137.1
NM_176796.2 909 Intron NP_789766.1
NM_176797.2 909 Intron NP_789767.1
NM_176798.2 909 Intron NP_789768.1
XM_005274022.3 909 Intron XP_005274079.1
XM_006718571.3 909 Intron XP_006718634.1
XM_011545076.2 909 UTR 5 XP_011543378.1
XM_011545077.2 909 UTR 5 XP_011543379.1
XM_011545079.2 909 Intron XP_011543381.1

View Full Product Details