Product Details

SNP ID
rs79815855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57038089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGATGAACCTCCAATTATTG[A/C]CCTACCATGCTCCAATATTGACCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5AK4P PubMed Links

Gene Details

Gene
OR5AK4P
Gene Name
olfactory receptor family 5 subfamily AK member 4 pseudogene
There are no transcripts associated with this gene.

View Full Product Details