Product Details

SNP ID
rs79989970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57407038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATGCCCAAAGTAAATAACTTA[C/T]ATCCCTTCTTGAAAACATCCCCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC43A3 PubMed Links

Gene Details

Gene
SLC43A3
Gene Name
solute carrier family 43 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278201.1 2562 UTR 3 NP_001265130.1
NM_001278206.1 2562 UTR 3 NP_001265135.1
NM_014096.3 2562 UTR 3 NP_054815.2
NM_017611.2 2562 UTR 3 NP_060081.1
NM_199329.2 2562 UTR 3 NP_955361.1

View Full Product Details