Product Details

SNP ID
rs77556759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85957675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCATCTCAGACCAGAATACCA[G/T]AACAAAACTTTTGTGGAAGCTGCAT
Phenotype
MIM: 603025
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PICALM PubMed Links

Gene Details

Gene
PICALM
Gene Name
phosphatidylinositol binding clathrin assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008660.2 3619 UTR 3 NP_001008660.1
NM_001206946.1 3619 UTR 3 NP_001193875.1
NM_001206947.1 3619 UTR 3 NP_001193876.1
NM_007166.3 3619 UTR 3 NP_009097.2
XM_005274322.2 3619 UTR 3 XP_005274379.1
XM_005274323.2 3619 UTR 3 XP_005274380.1
XM_005274324.2 3619 UTR 3 XP_005274381.1
XM_005274325.2 3619 UTR 3 XP_005274382.1
XM_005274326.2 3619 UTR 3 XP_005274383.1
XM_005274327.2 3619 UTR 3 XP_005274384.1
XM_005274328.2 3619 UTR 3 XP_005274385.1
XM_005274329.3 3619 UTR 3 XP_005274386.1
XM_005274330.2 3619 UTR 3 XP_005274387.1
XM_005274331.2 3619 UTR 3 XP_005274388.1
XM_005274332.2 3619 UTR 3 XP_005274389.1
XM_005274333.2 3619 UTR 3 XP_005274390.1
XM_005274334.2 3619 UTR 3 XP_005274391.1
XM_005274335.2 3619 UTR 3 XP_005274392.1
XM_005274336.2 3619 UTR 3 XP_005274393.1
XM_005274337.2 3619 UTR 3 XP_005274394.1
XM_005274340.2 3619 UTR 3 XP_005274397.1
XM_006718699.2 3619 UTR 3 XP_006718762.1
XM_006718700.2 3619 UTR 3 XP_006718763.1
XM_006718701.3 3619 Intron XP_006718764.1
XM_011545293.1 3619 UTR 3 XP_011543595.1
XM_017018381.1 3619 UTR 3 XP_016873870.1
XM_017018382.1 3619 UTR 3 XP_016873871.1
XM_017018383.1 3619 UTR 3 XP_016873872.1
XM_017018384.1 3619 UTR 3 XP_016873873.1
XM_017018385.1 3619 Intron XP_016873874.1
XM_017018386.1 3619 UTR 3 XP_016873875.1
XM_017018387.1 3619 UTR 3 XP_016873876.1
XM_017018388.1 3619 UTR 3 XP_016873877.1

View Full Product Details