Product Details

SNP ID
rs77337461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58708982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCACATCCCTGGGTCAAGGTGCT[C/T]CCACCTTCACCTTTTCATACACTCA
Phenotype
MIM: 607424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYAT PubMed Links

Gene Details

Gene
GLYAT
Gene Name
glycine-N-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005838.3 1893 Intron NP_005829.3
NM_201648.2 1893 UTR 3 NP_964011.2
XM_017017087.1 1893 UTR 3 XP_016872576.1

View Full Product Details