Product Details

SNP ID
rs79344328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59576232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAGATCTTGGCTGAGCAGGATA[C/G]TACAAGGGGAGGGTGAGTGACATTG
Phenotype
MIM: 167040
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OSBP PubMed Links

Gene Details

Gene
OSBP
Gene Name
oxysterol binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002556.2 3249 UTR 3 NP_002547.1

View Full Product Details