Product Details

SNP ID
rs76333679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60396115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTTGAGTTTGCCCCTTCCCTG[C/T]CCTGTATGTCTTTGGGTGACATAAC
Phenotype
MIM: 606502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A14 PubMed Links

Gene Details

Gene
MS4A14
Gene Name
membrane spanning 4-domains A14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079692.2 102 UTR 5 NP_001073160.1
NM_001261827.1 102 UTR 5 NP_001248756.1
NM_001261828.1 102 UTR 5 NP_001248757.1
NM_032597.4 102 UTR 5 NP_115986.3
Gene
MS4A7
Gene Name
membrane spanning 4-domains A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021201.4 102 Intron NP_067024.1
NM_206938.1 102 Intron NP_996821.1
NM_206939.1 102 Intron NP_996822.1
NM_206940.1 102 Intron NP_996823.1

View Full Product Details