Product Details

SNP ID
rs79270150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152168392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCTCAGTCGGGCTGTCCTGCA[T/C]GTAGGTGGCCTTGGGTGAAGCAATG
Phenotype
MIM: 305660
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GABRA3 PubMed Links

Gene Details

Gene
GABRA3
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000808.3 1343 Missense Mutation ATG,GTG M439V NP_000799.1
XM_006724811.2 1343 UTR 3 XP_006724874.1

View Full Product Details