Product Details

SNP ID
rs115587426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:234959525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATTCCTCCCTATGAATAATAAA[C/T]GGCTTACTTATAAATACTGTCCAGT
Phenotype
MIM: 605611
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3BP4 PubMed Links

Gene Details

Gene
SH3BP4
Gene Name
SH3 domain binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014521.2 Intron NP_055336.1
XM_011510891.2 Intron XP_011509193.1
XM_011510892.1 Intron XP_011509194.1
XM_011510893.1 Intron XP_011509195.1
XM_011510894.2 Intron XP_011509196.1

View Full Product Details