Product Details

SNP ID
rs113733302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170928557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAATACGACCTGGCTACCTGAA[A/G]GAGGTAGGACGGGGAACCGAGCAGC
Phenotype
MIM: 608693
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GORASP2 PubMed Links

Gene Details

Gene
GORASP2
Gene Name
golgi reassembly stacking protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201428.1 120 UTR 5 NP_001188357.1
NM_015530.4 120 UTR 5 NP_056345.3
XM_006712408.2 120 Intron XP_006712471.1

View Full Product Details