Product Details

SNP ID
rs116071964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:152122558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTACCAAACATAGCAAAATTT[C/T]ATATTTTATTCTAACTGAACACTAA
Phenotype
MIM: 606244
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STAM2 PubMed Links

Gene Details

Gene
STAM2
Gene Name
signal transducing adaptor molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005843.5 Intron NP_005834.4

View Full Product Details