Product Details

SNP ID
rs115896530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197572162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCATTCCCGGGAGGGAGAAAGG[C/T]GGCACTCCTCAGGCAGGTGTCTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFTN2 PubMed Links

Gene Details

Gene
RFTN2
Gene Name
raftlin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144629.2 1971 Missense Mutation CAC,CGC H451R NP_653230.2
XM_011510595.2 1971 Missense Mutation CAC,CGC H461R XP_011508897.1
XM_011510596.2 1971 Missense Mutation CAC,CGC H460R XP_011508898.1
XM_011510597.2 1971 Missense Mutation CAC,CGC H450R XP_011508899.1
XM_011510598.2 1971 Missense Mutation CAC,CGC H444R XP_011508900.1
XM_017003332.1 1971 Missense Mutation CAC,CGC H434R XP_016858821.1

View Full Product Details