Product Details

SNP ID
rs116491566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197705429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCTGCGAACGTCCGTCCTCCG[C/G]CTGCTAGGACGCACGGGGGCTAGTA
Phenotype
MIM: 609728
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MARS2 PubMed Links

Gene Details

Gene
MARS2
Gene Name
methionyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138395.3 126 Silent Mutation CGC,CGG R8R NP_612404.1

View Full Product Details