Product Details

SNP ID
rs112591281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219076447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCTGAGGTCTCTGCAGAGGGCC[G/T]GAAGTACCCTAGAAAAAAGGGAACC
Phenotype
MIM: 611290
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NHEJ1 PubMed Links

Gene Details

Gene
NHEJ1
Gene Name
non-homologous end joining factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024782.2 980 Silent Mutation TCA,TCC S278S NP_079058.1

View Full Product Details