Product Details

SNP ID
rs113556154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219502304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTGCATGCCAGGTGCTGTAG[C/T]GGAGGGAAAGAAAGAAAAAACAGAC
Phenotype
MIM: 615495 MIM: 615950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GMPPA PubMed Links

Gene Details

Gene
GMPPA
Gene Name
GDP-mannose pyrophosphorylase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013335.3 Intron NP_037467.2
NM_205847.2 Intron NP_995319.1
XM_005246483.3 Intron XP_005246540.1
XM_005246486.3 Intron XP_005246543.1
XM_011511032.2 Intron XP_011509334.1
XM_017003906.1 Intron XP_016859395.1
XM_017003907.1 Intron XP_016859396.1
XM_017003908.1 Intron XP_016859397.1
XM_017003909.1 Intron XP_016859398.1
XM_017003910.1 Intron XP_016859399.1
XM_017003911.1 Intron XP_016859400.1
XM_017003912.1 Intron XP_016859401.1
XM_017003913.1 Intron XP_016859402.1
XM_017003914.1 Intron XP_016859403.1
XM_017003915.1 Intron XP_016859404.1
XM_017003916.1 Intron XP_016859405.1
XM_017003917.1 Intron XP_016859406.1
Gene
LOC100996693
Gene Name
CAVP-target protein-like
There are no transcripts associated with this gene.

Gene
SPEG
Gene Name
SPEG complex locus
There are no transcripts associated with this gene.

View Full Product Details