Product Details

SNP ID
rs1804250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231712809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGTCTTTCTCTGCTTAGGATGA[C/T]GATGTCGATACCAAGAAGCAGAAGA
Phenotype
MIM: 188390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1244-1 PubMed Links

Gene Details

Gene
MIR1244-1
Gene Name
microRNA 1244-1
There are no transcripts associated with this gene.

Gene
PTMA
Gene Name
prothymosin, alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099285.1 Intron NP_001092755.1
NM_002823.4 Intron NP_002814.3

View Full Product Details