Product Details

SNP ID
rs114285297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232705276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATTAGAGTAGTAGTGGGGACC[A/G]TTACTTTTGGGAAAGGATCAGTGAG
Phenotype
MIM: 612003
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIGYF2 PubMed Links

Gene Details

Gene
GIGYF2
Gene Name
GRB10 interacting GYF protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103146.1 Intron NP_001096616.1
NM_001103147.1 Intron NP_001096617.1
NM_001103148.1 Intron NP_001096618.1
NM_015575.3 Intron NP_056390.2

View Full Product Details