Product Details
- SNP ID
-
rs112543214
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240560766 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCGCGTCGCGCCCTCACTCTTC[C/T]TCGGGAGCGCGCGAGCCGCGGGCGC
- Phenotype
-
MIM: 605287
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKMY1
PubMed Links
Gene Details
- Gene
- ANKMY1
- Gene Name
- ankyrin repeat and MYND domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282771.1 |
290 |
Intron |
|
|
NP_001269700.1 |
NM_001282780.1 |
290 |
Intron |
|
|
NP_001269709.1 |
NM_001282781.1 |
290 |
Intron |
|
|
NP_001269710.1 |
NM_001308375.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
NP_001295304.1 |
NM_016552.3 |
290 |
Intron |
|
|
NP_057636.2 |
NM_017844.3 |
290 |
Intron |
|
|
NP_060314.2 |
XM_011511300.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509602.1 |
XM_011511301.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509603.1 |
XM_011511304.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509606.1 |
XM_011511305.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509607.1 |
XM_011511309.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509611.1 |
XM_011511313.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509615.1 |
XM_011511315.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509617.1 |
XM_011511317.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509619.1 |
XM_011511318.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509620.1 |
XM_011511319.2 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509621.1 |
XM_011511322.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_011509624.1 |
XM_017004271.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859760.1 |
XM_017004272.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859761.1 |
XM_017004273.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859762.1 |
XM_017004274.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859763.1 |
XM_017004275.1 |
290 |
Intron |
|
|
XP_016859764.1 |
XM_017004276.1 |
290 |
UTR 5 |
|
|
XP_016859765.1 |
XM_017004277.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859766.1 |
XM_017004278.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859767.1 |
XM_017004279.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859768.1 |
XM_017004280.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859769.1 |
XM_017004281.1 |
290 |
Intron |
|
|
XP_016859770.1 |
XM_017004282.1 |
290 |
UTR 5 |
|
|
XP_016859771.1 |
XM_017004283.1 |
290 |
UTR 5 |
|
|
XP_016859772.1 |
XM_017004284.1 |
290 |
Missense Mutation |
AAG,AGG |
K62R |
XP_016859773.1 |
XM_017004285.1 |
290 |
Intron |
|
|
XP_016859774.1 |
XM_017004286.1 |
290 |
UTR 5 |
|
|
XP_016859775.1 |
XM_017004287.1 |
290 |
UTR 5 |
|
|
XP_016859776.1 |
XM_017004288.1 |
290 |
UTR 5 |
|
|
XP_016859777.1 |
XM_017004289.1 |
290 |
UTR 5 |
|
|
XP_016859778.1 |
XM_017004290.1 |
290 |
UTR 5 |
|
|
XP_016859779.1 |
XM_017004291.1 |
290 |
UTR 5 |
|
|
XP_016859780.1 |
- Gene
- DUSP28
- Gene Name
- dual specificity phosphatase 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033575.1 |
290 |
Missense Mutation |
CTC,TTC |
L28F |
NP_001028747.1 |
- Gene
- RNPEPL1
- Gene Name
- arginyl aminopeptidase like 1
There are no transcripts associated with this gene.
View Full Product Details