Product Details

SNP ID
rs114083698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240607144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGACATATTTCAATCTATGAAC[A/G]TGGTATACCCATCCATTTACTTATT
Phenotype
MIM: 605286 MIM: 602646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN10 PubMed Links

Gene Details

Gene
CAPN10
Gene Name
calpain 10
There are no transcripts associated with this gene.

Gene
GPR35
Gene Name
G protein-coupled receptor 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195381.1 Intron NP_001182310.1
NM_001195382.1 Intron NP_001182311.1
NM_005301.3 Intron NP_005292.2

View Full Product Details