Product Details

SNP ID
rs116715785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119431653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTAAAGATAATGAGGATTGAAA[C/G]AAAAGCAGACATACCAAAGGTACTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM39A PubMed Links

Gene Details

Gene
TMEM39A
Gene Name
transmembrane protein 39A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018266.2 2060 UTR 3 NP_060736.1
XM_005247578.1 2060 UTR 3 XP_005247635.1
XM_006713687.1 2060 UTR 3 XP_006713750.1
XM_011512958.2 2060 Intron XP_011511260.1
XM_017006788.1 2060 UTR 3 XP_016862277.1

View Full Product Details