Product Details

SNP ID
rs115645622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121667581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTCATTCACTTCCTGTCTGGTG[C/T]TGCATAGCTGCTGCTGAGCTTCAGA
Phenotype
MIM: 602500 MIM: 601306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLGB1 PubMed Links

Gene Details

Gene
GOLGB1
Gene Name
golgin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256486.1 9939 Missense Mutation AAC,AGC N3155S NP_001243415.1
NM_001256487.1 9939 Missense Mutation AAC,AGC N3111S NP_001243416.1
NM_001256488.1 9939 Missense Mutation AAC,AGC N3070S NP_001243417.1
NM_004487.4 9939 Missense Mutation AAC,AGC N3145S NP_004478.3
XM_005247371.4 9939 Missense Mutation AAC,AGC N3155S XP_005247428.1
XM_005247372.4 9939 Missense Mutation AAC,AGC N3116S XP_005247429.1
XM_005247373.1 9939 Missense Mutation AAC,AGC N3114S XP_005247430.1
XM_006713587.1 9939 Missense Mutation AAC,AGC N3150S XP_006713650.1
XM_006713588.1 9939 Missense Mutation AAC,AGC N3150S XP_006713651.1
XM_006713589.1 9939 Missense Mutation AAC,AGC N3149S XP_006713652.1
XM_006713590.1 9939 Missense Mutation AAC,AGC N3109S XP_006713653.1
XM_006713591.1 9939 Missense Mutation AAC,AGC N3075S XP_006713654.1
XM_011512699.2 9939 Missense Mutation AAC,AGC N3155S XP_011511001.1
XM_017006189.1 9939 Missense Mutation AAC,AGC N3150S XP_016861678.1
XM_017006190.1 9939 Missense Mutation AAC,AGC N3149S XP_016861679.1
XM_017006191.1 9939 Missense Mutation AAC,AGC N3114S XP_016861680.1
XM_017006192.1 9939 Missense Mutation AAC,AGC N3111S XP_016861681.1
XM_017006193.1 9939 Missense Mutation AAC,AGC N3110S XP_016861682.1
XM_017006194.1 9939 Missense Mutation AAC,AGC N3075S XP_016861683.1
XM_017006195.1 9939 Missense Mutation AAC,AGC N2902S XP_016861684.1
Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
There are no transcripts associated with this gene.

View Full Product Details