Product Details

SNP ID
rs114373610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131028037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATTTATTTGAACAGCTTAGTAT[A/G]GCCAGACGGCATCACTTTTATGACA
Phenotype
MIM: 609779
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASTE1 PubMed Links

Gene Details

Gene
ASTE1
Gene Name
asteroid homolog 1 (Drosophila)
There are no transcripts associated with this gene.

Gene
NEK11
Gene Name
NIMA related kinase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146003.1 Intron NP_001139475.1
NM_001321220.1 Intron NP_001308149.1
NM_001321221.1 Intron NP_001308150.1
NM_001321222.1 Intron NP_001308151.1
NM_001321223.1 Intron NP_001308152.1
NM_001321224.1 Intron NP_001308153.1
NM_024800.4 Intron NP_079076.3
NM_145910.3 Intron NP_665917.1
XM_011513167.2 Intron XP_011511469.1
XM_011513173.1 Intron XP_011511475.1
XM_011513174.1 Intron XP_011511476.1
XM_011513175.1 Intron XP_011511477.1
XM_011513181.1 Intron XP_011511483.1
XM_017007205.1 Intron XP_016862694.1
XM_017007206.1 Intron XP_016862695.1
XM_017007207.1 Intron XP_016862696.1
XM_017007208.1 Intron XP_016862697.1
XM_017007209.1 Intron XP_016862698.1
XM_017007210.1 Intron XP_016862699.1
XM_017007211.1 Intron XP_016862700.1
XM_017007212.1 Intron XP_016862701.1
XM_017007213.1 Intron XP_016862702.1
XM_017007214.1 Intron XP_016862703.1
XM_017007215.1 Intron XP_016862704.1
XM_017007216.1 Intron XP_016862705.1
XM_017007217.1 Intron XP_016862706.1
XM_017007218.1 Intron XP_016862707.1
XM_017007219.1 Intron XP_016862708.1
XM_017007220.1 Intron XP_016862709.1
XM_017007221.1 Intron XP_016862710.1
XM_017007222.1 Intron XP_016862711.1
XM_017007223.1 Intron XP_016862712.1
XM_017007224.1 Intron XP_016862713.1
XM_017007225.1 Intron XP_016862714.1
XM_017007226.1 Intron XP_016862715.1
XM_017007227.1 Intron XP_016862716.1
XM_017007228.1 Intron XP_016862717.1
XM_017007229.1 Intron XP_016862718.1
XM_017007230.1 Intron XP_016862719.1
XM_017007231.1 Intron XP_016862720.1
XM_017007232.1 Intron XP_016862721.1
XM_017007233.1 Intron XP_016862722.1

View Full Product Details