Product Details
- SNP ID
-
rs116617551
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:133674610 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGCCAGCCACGGAGAATGAGGCT[C/T]GCTATCTGCGCCCTGCTGTGCGCCG
- Phenotype
-
MIM: 190000
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TF
PubMed Links
Gene Details
- Gene
- TF
- Gene Name
- transferrin
View Full Product Details