Product Details

SNP ID
rs115085348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134357266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTAGCCAGAGAACAATATGGCTG[A/G]CTTGATTAGTGAACCAGTCCACCTG
Phenotype
MIM: 614658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMOTL2 PubMed Links

Gene Details

Gene
AMOTL2
Gene Name
angiomotin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278683.1 2952 UTR 3 NP_001265612.1
NM_001278685.1 2952 UTR 3 NP_001265614.1
NM_016201.3 2952 UTR 3 NP_057285.3
XM_006713654.2 2952 UTR 3 XP_006713717.1
XM_011512881.1 2952 UTR 3 XP_011511183.1
XM_017006580.1 2952 UTR 3 XP_016862069.1
XM_017006581.1 2952 UTR 3 XP_016862070.1
XM_017006582.1 2952 UTR 3 XP_016862071.1
Gene
RPL39P5
Gene Name
ribosomal protein L39 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details