Product Details

SNP ID
rs115218832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136337492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAATGCACTGTATTTGAATCTC[C/T]CTAGTCTATATAAAATGAACGGTGT
Phenotype
MIM: 232050 MIM: 604358
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCCB PubMed Links

Gene Details

Gene
PCCB
Gene Name
propionyl-CoA carboxylase beta subunit
There are no transcripts associated with this gene.

Gene
STAG1
Gene Name
stromal antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005862.2 3403 UTR 3 NP_005853.2
XM_011512329.2 3403 UTR 3 XP_011510631.1
XM_011512331.2 3403 UTR 3 XP_011510633.1
XM_017005523.1 3403 UTR 3 XP_016861012.1
XM_017005524.1 3403 UTR 3 XP_016861013.1
XM_017005525.1 3403 UTR 3 XP_016861014.1

View Full Product Details