Product Details

SNP ID
rs117725617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:137764934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTGCCGAATGGAAGCTTCTGTC[C/T]GAGGCAGAGAAGCGGCCATACATCG
Phenotype
MIM: 604747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01210 PubMed Links

Gene Details

Gene
LINC01210
Gene Name
long intergenic non-protein coding RNA 1210
There are no transcripts associated with this gene.

Gene
SOX14
Gene Name
SRY-box 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004189.3 643 Silent Mutation TCC,TCT S50S NP_004180.1

View Full Product Details