Product Details
- SNP ID
-
rs111721329
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139940394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTGAGACTCAGTTTCCTCATCT[A/C]TAAAATGGGGATACAGAGATCTACC
- Phenotype
-
MIM: 611323
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLSTN2
PubMed Links
Gene Details
- Gene
- CLSTN2
- Gene Name
- calsyntenin 2
View Full Product Details