Product Details

SNP ID
rs115729997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156675425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGTAAAGAAGAATTAGAGAGA[A/G]AAGGCGCCGCGAGTAGTCTCTGGGA
Phenotype
MIM: 612480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIPARP PubMed Links

Gene Details

Gene
TIPARP
Gene Name
TCDD inducible poly(ADP-ribose) polymerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184717.1 Intron NP_001171646.1
NM_001184718.1 Intron NP_001171647.1
NM_015508.4 Intron NP_056323.2
Gene
TIPARP-AS1
Gene Name
TIPARP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details