Product Details

SNP ID
rs113278243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101229402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTCTCTGGAAAGCTCACTGCTC[C/T]CATACATCTGTCTGATTTCTTCTCT
Phenotype
MIM: 607056
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPG2 PubMed Links

Gene Details

Gene
IMPG2
Gene Name
interphotoreceptor matrix proteoglycan 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016247.3 3814 Missense Mutation GAG,GGG E1204G NP_057331.2

View Full Product Details