Product Details
- SNP ID
-
rs112537671
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:101327733 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCACTGCTATTGTCCTGTTTAGGA[A/G]CTTTAGGGCATAGATCCACCATGTT
- Phenotype
-
MIM: 607056
MIM: 612846
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IMPG2
PubMed Links
Gene Details
- Gene
- IMPG2
- Gene Name
- interphotoreceptor matrix proteoglycan 2
There are no transcripts associated with this gene.
- Gene
- SENP7
- Gene Name
- SUMO1/sentrin specific peptidase 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001077203.2 |
3048 |
Missense Mutation |
GCT,GTT |
A918V |
NP_001070671.1 |
NM_001282801.1 |
3048 |
Missense Mutation |
GCT,GTT |
A917V |
NP_001269730.1 |
NM_001282802.1 |
3048 |
Missense Mutation |
GCT,GTT |
A950V |
NP_001269731.1 |
NM_001282803.1 |
3048 |
Missense Mutation |
GCT,GTT |
A819V |
NP_001269732.1 |
NM_001282804.1 |
3048 |
Intron |
|
|
NP_001269733.1 |
NM_020654.4 |
3048 |
Missense Mutation |
GCT,GTT |
A983V |
NP_065705.3 |
XM_005247662.4 |
3048 |
Missense Mutation |
GCT,GTT |
A852V |
XP_005247719.1 |
XM_011513038.2 |
3048 |
Missense Mutation |
GCT,GTT |
A984V |
XP_011511340.1 |
XM_011513039.2 |
3048 |
Missense Mutation |
GCT,GTT |
A951V |
XP_011511341.1 |
XM_011513040.2 |
3048 |
Missense Mutation |
GCT,GTT |
A919V |
XP_011511342.1 |
XM_011513041.2 |
3048 |
Missense Mutation |
GCT,GTT |
A918V |
XP_011511343.1 |
XM_011513042.2 |
3048 |
Missense Mutation |
GCT,GTT |
A853V |
XP_011511344.1 |
XM_011513045.2 |
3048 |
Intron |
|
|
XP_011511347.1 |
XM_017006924.1 |
3048 |
Missense Mutation |
GCT,GTT |
A951V |
XP_016862413.1 |
XM_017006925.1 |
3048 |
Missense Mutation |
GCT,GTT |
A950V |
XP_016862414.1 |
XM_017006926.1 |
3048 |
Missense Mutation |
GCT,GTT |
A885V |
XP_016862415.1 |
XM_017006927.1 |
3048 |
Missense Mutation |
GCT,GTT |
A884V |
XP_016862416.1 |
XM_017006928.1 |
3048 |
Intron |
|
|
XP_016862417.1 |
XM_017006929.1 |
3048 |
Intron |
|
|
XP_016862418.1 |
View Full Product Details