Product Details

SNP ID
rs116773206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172632353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAAGGCAGTTTTTTAAAAAATA[C/T]CATTGTTTTTAATATTTATTATGGA
Phenotype
MIM: 613234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCEH1 PubMed Links

Gene Details

Gene
NCEH1
Gene Name
neutral cholesterol ester hydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146276.1 2583 UTR 3 NP_001139748.1
NM_001146277.1 2583 UTR 3 NP_001139749.1
NM_001146278.1 2583 UTR 3 NP_001139750.1
NM_020792.4 2583 UTR 3 NP_065843.3

View Full Product Details