Product Details

SNP ID
rs116044363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179328742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGACTTTATCCTCTTTATTACTT[C/T]AGGAAATGTAAACTTTCATGCTGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF639 PubMed Links

Gene Details

Gene
ZNF639
Gene Name
zinc finger protein 639
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303425.1 Intron NP_001290354.1
NM_001303426.1 Intron NP_001290355.1
NM_016331.2 Intron NP_057415.1
XM_017006550.1 Intron XP_016862039.1
XM_017006551.1 Intron XP_016862040.1
XM_017006552.1 Intron XP_016862041.1
XM_017006553.1 Intron XP_016862042.1

View Full Product Details