Product Details

SNP ID
rs114192437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:183833016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACAGCGGGTACTCAGTAAGTGG[C/T]CCACATTGGTTTGCAGTCACCGGCC
Phenotype
MIM: 610593 MIM: 607858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP6D1 PubMed Links

Gene Details

Gene
MAP6D1
Gene Name
MAP6 domain containing 1
There are no transcripts associated with this gene.

Gene
PARL
Gene Name
presenilin associated rhomboid like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037639.2 Intron NP_001032728.1
NM_001324436.1 Intron NP_001311365.1
NM_001324437.1 Intron NP_001311366.1
NM_001324438.1 Intron NP_001311367.1
NM_018622.6 Intron NP_061092.3
XM_005247582.4 Intron XP_005247639.1
XM_017006800.1 Intron XP_016862289.1
XM_017006801.1 Intron XP_016862290.1
XM_017006802.1 Intron XP_016862291.1
XM_017006803.1 Intron XP_016862292.1

View Full Product Details