Product Details

SNP ID
rs112656750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186639296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCATGAGGGTGGAGCCCTCATCA[A/G]GATAGGATAATCCTCTCAGCATTAC
Phenotype
MIM: 605954
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FETUB PubMed Links

Gene Details

Gene
FETUB
Gene Name
fetuin B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308077.1 Intron NP_001295006.1
NM_001308079.1 Intron NP_001295008.1
NM_014375.2 Intron NP_055190.2
XM_005247351.3 Intron XP_005247408.1
XM_011512681.2 Intron XP_011510983.1
XM_011512682.2 Intron XP_011510984.1
XM_011512683.2 Intron XP_011510985.1

View Full Product Details