Product Details

SNP ID
rs112108518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:8270326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCTCCGGGACGCCCGTGCGC[A/C]AGCTGCAGAAGGGCGCCTGCCCGTT
Phenotype
MIM: 608785
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HTRA3 PubMed Links

Gene Details

Gene
HTRA3
Gene Name
HtrA serine peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297559.1 570 Missense Mutation AAG,CAG K120Q NP_001284488.1
NM_053044.4 570 Missense Mutation AAG,CAG K120Q NP_444272.1
XM_011513596.2 570 Missense Mutation AAG,CAG K120Q XP_011511898.1

View Full Product Details