Product Details

SNP ID
rs115460634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42409264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCATACAGAAAGTAAATCAAGA[C/T]GAAATATGGAAAACATTTATTTCCT
Phenotype
MIM: 609542
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8A1 PubMed Links

Gene Details

Gene
ATP8A1
Gene Name
ATPase phospholipid transporting 8A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105529.1 7334 UTR 3 NP_001098999.1
NM_006095.2 7334 UTR 3 NP_006086.1
XM_005248043.2 7334 UTR 3 XP_005248100.1
XM_011513615.1 7334 UTR 3 XP_011511917.1
XM_011513616.2 7334 UTR 3 XP_011511918.1
XM_011513618.1 7334 UTR 3 XP_011511920.1
XM_017007645.1 7334 UTR 3 XP_016863134.1
XM_017007646.1 7334 UTR 3 XP_016863135.1
XM_017007647.1 7334 UTR 3 XP_016863136.1
Gene
SHISA3
Gene Name
shisa family member 3
There are no transcripts associated with this gene.

View Full Product Details