Product Details
- SNP ID
-
rs114448516
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:1321204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAACTCAGCTCCGCCTGCAATGC[C/T]GAGGCACTTCTAATCCCACACGGAG
- Phenotype
-
MIM: 612585
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLPTM1L
PubMed Links
Gene Details
- Gene
- CLPTM1L
- Gene Name
- CLPTM1 like
View Full Product Details