Product Details

SNP ID
rs116132124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37815720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAACCAGGTTATCATCTAAAAA[C/T]GACAGGTCATCATCAAAGGCGATGG
Phenotype
MIM: 600837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDNF PubMed Links

Gene Details

Gene
GDNF
Gene Name
glial cell derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000514.3 787 Silent Mutation TCA,TCG S189S NP_000505.1
NM_001190468.1 787 Silent Mutation TCA,TCG S206S NP_001177397.1
NM_001190469.1 787 Silent Mutation TCA,TCG S180S NP_001177398.1
NM_001278098.1 787 Silent Mutation TCA,TCG S137S NP_001265027.1
NM_199231.2 787 Silent Mutation TCA,TCG S163S NP_954701.1
XM_011514030.2 787 Silent Mutation TCA,TCG S137S XP_011512332.1
XM_017009337.1 787 Silent Mutation TCA,TCG S163S XP_016864826.1

View Full Product Details