Product Details

SNP ID
rs112350718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54979371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATTTCCTCATTACGGGAGACCC[A/G]GCAGCATTCTCTTTCACAACTTCTT
Phenotype
MIM: 601521
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ESM1 PubMed Links

Gene Details

Gene
ESM1
Gene Name
endothelial cell specific molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135604.1 585 Silent Mutation GCC,GCT A122A NP_001129076.1
NM_007036.4 585 Silent Mutation GCC,GCT A172A NP_008967.1

View Full Product Details