Product Details

SNP ID
rs114683330
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:75254581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGTGGCACATTTCTACTATTAA[C/G]CATTACATTGTTTTTTGTTTTTTAA
Phenotype
MIM: 611028
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
TMEM30A PubMed Links

Gene Details

Gene
TMEM30A
Gene Name
transmembrane protein 30A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143958.1 2777 UTR 3 NP_001137430.1
NM_018247.3 2777 UTR 3 NP_060717.1

View Full Product Details