Product Details

SNP ID
rs114491179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11102570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAAGCAACTGGGTCTCTACTA[C/T]AGAGGTACAAGGGGGAAAATTTTAT
Phenotype
MIM: 610524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERVFRD-1 PubMed Links

Gene Details

Gene
ERVFRD-1
Gene Name
endogenous retrovirus group FRD member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207582.2 3223 UTR 3 NP_997465.1
Gene
SMIM13
Gene Name
small integral membrane protein 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135575.1 3223 Intron NP_001129047.1

View Full Product Details