Product Details

SNP ID
rs114654366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:18390442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTAAATGAGTAAAAATGACCAA[C/T]GCTTCTGCAACGTGGAATGAGGATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF144B PubMed Links

Gene Details

Gene
RNF144B
Gene Name
ring finger protein 144B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182757.3 Intron NP_877434.2
XM_005248986.3 Intron XP_005249043.1

View Full Product Details