Product Details

SNP ID
rs114711913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:20101763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGCCACACCACTCAAATGTCCTA[C/T]AAAATATCAAAACCAATAACCTGGA
Phenotype
MIM: 611732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBOAT1 PubMed Links

Gene Details

Gene
MBOAT1
Gene Name
membrane bound O-acyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080480.2 1978 UTR 3 NP_001073949.1
XM_006714999.2 1978 UTR 3 XP_006715062.1
XM_006715000.3 1978 Intron XP_006715063.1
XM_011514313.2 1978 Intron XP_011512615.1

View Full Product Details