Product Details
- SNP ID
-
rs112921331
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:22570091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCCGACCAAGAGGAGGAGGCGGA[A/G]GCGGAGAGGGCGGCGGAAGCGGAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HDGFL1
PubMed Links
Gene Details
- Gene
- HDGFL1
- Gene Name
- hepatoma derived growth factor-like 1
There are no transcripts associated with this gene.
View Full Product Details