Product Details

SNP ID
rs113783901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29828736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGGCGGCCAATGTGGCTGAACA[A/G]AGGAGAGCCTACCTGGAGGGCACGT
Phenotype
MIM: 142871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-G PubMed Links

Gene Details

Gene
HLA-G
Gene Name
major histocompatibility complex, class I, G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002127.5 624 Silent Mutation CAA,CAG Q179Q NP_002118.1
XM_017010816.1 624 Silent Mutation CAA,CAG Q184Q XP_016866305.1
XM_017010817.1 624 Intron XP_016866306.1
XM_017010818.1 624 Intron XP_016866307.1

View Full Product Details